|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | FOXP3 |
Antigen Name | Forkhead Box P3 |
Reactivity | Mouse |
Conjugation | PE (Phycoerythrin) |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | MF23 |
Host | Rat |
Isotype | IgG2b |
Flow Cytometry | Yes |
Antigen Synonyms | DIETER, PIDX, AIID, JM2, Scurfin, IPEX, XPID |
Concentration | 0.2 mg/ml |
Cross Adsorption | No |
Purification | purified utilizing affinity chromatography and unreacted dye was removed from the product. |
Form | liquid |
Storage Buffer | Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. |
Storage Temperature | The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze. |
Shipping Temperature | -20°C |