Anti-ENG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: STA]

Supplier: ProSci
76-884
10765-798EA 340.73 USD
10765-798
Anti-ENG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: STA]
Antibodies
The STA monoclonal antibody specifically reacts with human CD106, a 110 kDA glycoprotein also named Vascular Cell Adhesion Molecule-1 (VCAM-1) and INCAM-110. Its expression is induced by inflammation stimuli and cytokines and it is found mainly on activated vascular endothelium. CD106 is also found on subsets of macrophages, stromal cells, dendritic cells, and myeloid lineage cells. The integrins that bind CD106 are CD49d/CD29 and LPAM-1, and it is involved in cell adhesion, transmigration and T cell proliferation.

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: STA
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR