Anti-CD3E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHT1]

Supplier: ProSci
76-290
10764-308EA 245.14 USD
10764-308
Anti-CD3E Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: UCHT1]
Antibodies
The UCHT1 monoclonal antibody specifically reacts with the epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains gamma, delta, and epsilon chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. Unlike HIT3a, another specific antibody of CD3, the UCHT1 antibody can stain both the surface and intracellular CD3 epsilon. The immobilized UCHT1 can cross-link with the TCR complex, enhancing cellular activation and proliferation.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR