Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3E Hamster Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 145-2C11]

Supplier: Prosci
76-276
The 145-2C11 monoclonal antibody reacts with mouse CD3e, the 20 kDa epsilon chain of the TCR complex. Together with the gamma and delta subunits of CD3, the epsilon subunits are involved in the assembly, trafficking, and surface expression of T-cell receptor complex. CD3 is expressed on thymocytes, mature T cells, and natural killer T cells, and the epsilon chain enhances the antigen recognition.The 145-2C11 antibody binds to the TCR complex and, depending on the conditions, initiates T cells activation, proliferation, and apoptosis. The soluble antibody seems to block lysis of target cells by antigen-specific cytotoxic T lymphocytes. The 145-2C11 antibody does not cross-react with the rat leukocytes, and it is used as a phenotypic marker for mouse T lymphocytes.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 µg 76-276 10764-304 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CD3E
Antigen Name CD3 Molecule Epsilon
Reactivity Mouse
Conjugation PE (Phycoerythrin)-Cy7®
Clonality Monoclonal
Antibody Type Primary
Clone 145-2C11
Host Hamster
Isotype IgG
Flow Cytometry Yes
Antigen Synonyms T-cell surface glycoprotein CD3 epsilon chain, TCRE, FLJ18683, T3E, T-cell surface antigen T3/Leu-4 epsilon chain, CD3e antigen, epsilon subunit of T3, CD3-epsilon, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex
Concentration 0.2 mg/ml
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty