Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]

Supplier: Prosci
The 10F.9G2 monoclonal antibody specifically reacts with mouse CD274, also known as B7-H1 or PD-L1, a 43 kDa glycoprotein of the B7 family of the immunoglobulin superfamily. CD274 is expressed on the B and T lymphocytes, natural killer cells, and dendritic cells. The receptor for the B7-H1 molecule is PD-1, which contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM), and is expressed on activated B and T cells. The interaction between CD274 and PD-1 seems to downregulate the T and B immune responses.

Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 10F.9G2
Conjugation: Unconjugated
Host: Rat
Isotype: IgG1
Reactivity: Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 µg 76-959 10765-820 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol CD274
Antigen Name CD274 molecule
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 10F.9G2
Host Rat
Isotype IgG2b kappa
Flow Cytometry Yes
Antigen Synonyms B7-H, PDCD1LG1, PDL1, PDCD1L1, B7H1, PD-L1
Concentration 2 mg/ml
Format purified antibody
Cross Adsorption No
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty