Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMP1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H4A3]

Supplier: Prosci
76-890
The H4A3 monoclonal antibody specifically reacts with human CD107a, a heavily glycosylated type I membrane glycoprotein. CD107a is also known as Lysosomal-associated membrane protein 1 (LAMP-1) and is widely expressed intracellular antigen. It can be found on the surface of PHA-activated lymphocytes, activated platelets, cytotoxic T cells, NK cells, macrophages, epithelial cells, endothelial cells, and some tumor lines. CD107a is a ligand for E-selectin and galaptin and is reported to be involved in cell adhesion and tumor metastasis.

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: H4A3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 tests 76-890 10765-810 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol LAMP1
Antigen Name Lysosomal-associated membrane protein 1
Reactivity Human
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Monoclonal
Antibody Type Primary
Clone H4A3
Host Mouse
Isotype IgG1 kappa
Flow Cytometry Yes
Antigen Synonyms LGP120, LAMPA, Lysosome-associated membrane glycoprotein 1, CD107 antigen-like family member A, LAMP-1, CD107a
Concentration 0.5 mg/ml
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty