|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | LAMP1 |
Antigen Name | Lysosomal-associated membrane protein 1 |
Reactivity | Human |
Conjugation | FITC (Fluorescein Isothiocyanate) |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | H4A3 |
Host | Mouse |
Isotype | IgG1 kappa |
Flow Cytometry | Yes |
Antigen Synonyms | LGP120, Lysosome-associated membrane glycoprotein 1, LAMP-1, LAMPA, CD107 antigen-like family member A, CD107a |
Concentration | 0.5 mg/ml |
Cross Adsorption | No |
Purification | purified utilizing affinity chromatography and unreacted dye was removed from the product. |
Form | liquid |
Storage Buffer | Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. |
Storage Temperature | The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze. |
Shipping Temperature | -20°C |