Anti-Csf1r Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AFS98]

Supplier: ProSci
76-878
10765-786EA 345.65 USD
10765-786
Anti-Csf1r Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AFS98]
Antibodies
The AFS98 monoclonal antibody specifically reacts with the mouse CD115 molecule, a 150 kDa receptor for the colony stimulating factor (CSF-1) or macrophage CFS (M-CFS), expressed by some epithelial cells, monocytes, osteoclasts, and macrophages. The colony stimulating factor-1 regulates the proliferation and the differentiation of the monocytic lineage cells. The AFS98 antibody can be used to identify myeloid lineage cells.

Type: Primary
Antigen: Csf1r
Clonality: Monoclonal
Clone: AFS98
Conjugation: FITC
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR