Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR4 Rat Monoclonal Antibody [clone: MTS510]

Supplier: Prosci
The MTS510 monoclonal antibody specifically reacts with the Toll-Like Receptor 4 (TLR-4) and MD-2 (LY96) molecular complex. The complex of TLR-4, MD-2, and CD14 regulates the innate immune system recognition of bacterial lipopolysaccharides (LPS) and is expressed on the surface of thioglycollate-elicited macrophages. The epitope that binds the MTS510 antibody is lost after LPS stimulation and the antibody can be used to co-immunoprecipitates MD-2 and TLR4.

Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: MTS510
Conjugation: Unconjugated
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 77-016 10765-260 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol TLR4
Antigen Name Toll-like Receptor 4
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone MTS510
Host Rat
Isotype IgG2a kappa
Flow Cytometry Yes
Antigen Synonyms Lps, TLR-4, Toll-like receptor 4, CD284, TOLL, hToll, TLR4, Ly87, Rasl2-8, ARMD10, Ran/M1
Concentration 1 mg/ml
Format purified antibody
Cross Adsorption No
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty