|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | JAG1 |
Antigen Name | Jagged 1 |
Reactivity | Mouse |
Conjugation | PE (Phycoerythrin) |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | HMJ1-29 |
Host | Hamster |
Isotype | IgG |
Flow Cytometry | Yes |
Antigen Synonyms | ABE2, Htu, Jag1, JAGL1, CD339, AHD, AWS, Gsfabe2, Ser-1, Ozz, jagged, AGS, HJ1 |
Concentration | 0.2 mg/ml |
Cross Adsorption | No |
Purification | purified utilizing affinity chromatography and unreacted dye was removed from the product. |
Form | liquid |
Storage Buffer | Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. |
Storage Temperature | The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze. |
Shipping Temperature | -20°C |