Anti-HLA-DRA Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5®.5) [clone: LN3]

Supplier: ProSci
77-013
10765-254EA 469.76 USD
10765-254
Anti-HLA-DRA Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5®.5) [clone: LN3]
Antibodies
The LN3 monoclonal antibody specifically reacts with the human leukocyte antigen-DR (HLA-DR), a MHC class II heterodimeric surface glycoprotein. HLA-DR is expressed on the antigen presenting surfaces of monocytes, macrophages, dendritic cells, activated T cells, and B cells. It is integral to peptide presentation to CD4+ T cells.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: LN3
Conjugation: PerCP-Cy5.5
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR