Anti-CD83 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB15e]

Supplier: ProSci
76-315
10764-358EA 683.4 USD
10764-358
Anti-CD83 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB15e]
Antibodies
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR