Anti-NT5E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AD2]

Supplier: ProSci
76-311
10764-350EA 717.56 USD
10764-350
Anti-NT5E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AD2]
Antibodies
Type: Primary
Antigen: NT5E
Clonality: Monoclonal
Clone: AD2
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR