Anti-CD33 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: WM53]

Supplier: ProSci
76-309
10764-346EA 491.73 USD
10764-346
Anti-CD33 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: WM53]
Antibodies
The WM53 monoclonal antibody specifically reacts with human CD33, a 67 kDA transmembrane glycoprotein also known as GP67 and P67. CD33 is expressed on monocytes, granulocytes, dendritic cells, mast cells, and myeloid progenitors to the exclusion of hematopoietic stem cells, lymphocytes, and platelets. It functions to assist sialic acid-dependent cell adhesion.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM53
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR