Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CEACAM8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: G10F5]

Supplier: Prosci
76-594
The G10F5 monoclonal antibody specifically reacts with human CD66b, a 100kDa glycosylphosphatidylinositol (GPI) linked protein in the granulocyte-specific activation family. CD66b is expressed on granulocytes, neutrophils, and eosinophils. It is reported that this molecule plays a role in regulating neutrophil activation and cellular adhesion. It was previously classified as CD67, but renamed CD66b in the Fifth HLDA Workshop.

Type: Primary
Antigen: CEACAM8
Clonality: Monoclonal
Clone: G10F5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 tests 76-594 10764-696 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CEACAM8
Antigen Name Carcinoembryonic Antigen-related Cell Adhesion Molecule 7
Reactivity Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody Type Primary
Clone G10F5
Host Mouse
Isotype IgM kappa
Flow Cytometry Yes
Antigen Synonyms NCA-95, CEACAM8, CGM6, CD66b, CD67
Concentration 0.2 mg/ml
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty