Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3]

Supplier: ProSci
76-385
10764-450EA 603.71 USD
10764-450
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3]
Antibodies
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR