Anti-ITGB1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HMb1-1]

Supplier: ProSci
76-796
10764-860EA 491.73 USD
10764-860
Anti-ITGB1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HMb1-1]
Antibodies
The HMb1-1 monoclonal antibody specifically reacts with mouse/rat CD29, a 130 kDA molecule also known as integrin beta 1, GPIIa, and the VLA-beta chain. It is expressed broadly on endothelial cells, epithelial, leukocytes, and smooth muscle. CD29 forms the VLA 1-6 complexes through the non-covalent interaction with the alpha integrins of CD49 a-f. The HMb1-1 antibody is capable of inhibiting T cell proliferation and blocking cell adhesion.

Type: Primary
Antigen: Itgb1
Clonality: Monoclonal
Clone: HMb1-1
Conjugation: PE
Epitope:
Host: Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR