Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]

Supplier: ProSci
76-336
10766-404EA 535.13 USD
10766-404
Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]
Antibodies
The GK1.5 monoclonal antibody specifically binds with the mouse CD4 molecule, also known as L3T4, a 55 kDa differentiation antigen which binds to the MHC class II. CD4 is expressed on most thymocytes, a subpopulation of mature T lymphocytes, dendritic cells, pluripotent hematopoietic stem cells, B cell precursors, and lymphoid precursors inside the thymus. It is also expressed on the mouse egg cell membrane, enhancing adhesion to MHC class II bearing sperm. By interaction with MHC class II on the surface of APC, CD4 initiates the development of T lymphocytes and helps the optimum functioning of mature T lymphocytes. The binding of the GK1.5 antibody blocks the binding of the Anti-Mouse CD4 RM4-5 antibody.

Type: Primary
Antigen: Cd4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR