Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD80 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2D10.4]

Supplier: Prosci
76-038
The 2D10.4 antibody reacts with human CD80, also known as B7-1, a 55 kDa type I transmembrane protein ligand for CD152 (CTLA-4) and for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 also binds a second B7 ligand known as CD86 (B7-2). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 or CTLA-4 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 2D10.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 Tests 76-038 10766-176 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CD80
Antigen Name CD80 molecule
Reactivity Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody Type Primary
Clone 2D10.4
Host Mouse
Isotype IgG1 kappa
Flow Cytometry Yes
Antigen Synonyms B71, MIC17, Cd28l, Ly-53, Ly53, Cluster of Differentiation 80, TSA1
Concentration 5 uL (1 ug) / test
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty