Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 104]

Supplier: ProSci
76-556
10764-652EA 448.24 USD
10764-652
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 104]
Antibodies
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 104
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR