Anti-FOXP3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3G3]

Supplier: ProSci
77-147
10765-986EA 535.12 USD
10765-986
Anti-FOXP3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3G3]
Antibodies
Type: Primary
Antigen: Foxp3
Clonality: Monoclonal
Clone: 3G3
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR