Anti-CD69 Hamster Monoclonal Antibody [clone: H1.2F3]

Supplier: ProSci
76-818
10764-888EA 345.65 USD
10764-888
Anti-CD69 Hamster Monoclonal Antibody [clone: H1.2F3]
Antibodies
The H1.2F3 monoclonal antibody specifically reacts with human CD69, the 27-33 kDA type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: Cd69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR