Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGB1 Hamster Monoclonal Antibody [clone: HMb1-1]

Supplier: Prosci
76-795
The HMb1-1 monoclonal antibody specifically reacts with mouse/rat CD29, a 130 kDA molecule also known as integrin beta 1, GPIIa, and the VLA-beta chain. It is expressed broadly on endothelial cells, epithelial, leukocytes, and smooth muscle. CD29 forms the VLA 1-6 complexes through the non-covalent interaction with the alpha integrins of CD49 a-f. The HMb1-1 antibody is capable of inhibiting T cell proliferation and blocking cell adhesion.

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: HMb1-1
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 µg 76-795 10764-858 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ITGB1
Antigen Name Integrin beta 1
Reactivity Mouse, Rat
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone HMb1-1
Host Hamster
Isotype IgG
Flow Cytometry Yes
Antigen Synonyms VLA-4 subunit beta, VLAB, VLA-BETA, GPIIA, MSK12, beta polypeptide, Fibronectin receptor subunit beta, integrin VLA-4 beta subunit, FNRB, CD29, MDF2, very late activation protein
Concentration 1 mg/ml
Format purified antibody
Cross Adsorption No
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty