Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: TS2/16]

Supplier: Prosci
The TS2/16 moncolonal antibody specifically reacts with human CD29 (Integrin beta1), a 130 kDA type I glycoprotein expressed on hematopoietic and non-hematopoietic cells. It forms the VLA-(1-6) molecules with integrin alpha-(1-6) and is involved in the adhesion between cell-cell and cell-matrix. The TS2/16 antibody has been found to activate beta 1 integrins.

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: TS2/16
Conjugation: PE (Phycoerythrin)
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 tests 76-793 10764-854 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol ITGB1
Antigen Name Integrin beta 1
Reactivity Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody Type Primary
Clone TS2/16
Host Mouse
Isotype IgG1 kappa
Flow Cytometry Yes
Antigen Synonyms VLA-BETA, MDF2, CD29, GPIIA, integrin VLA-4 beta subunit, VLAB, beta polypeptide, Fibronectin receptor subunit beta, very late activation protein, MSK12, FNRB, VLA-4 subunit beta
Concentration 20 uL (0.25 ug) / test
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty