Anti-ITGA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9F10]

Supplier: ProSci
76-803
10764-874EA 553.09 USD
10764-874
Anti-ITGA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9F10]
Antibodies
The 9F10 monoclonal antibody specifically reacts with human CD49d, the 150 kDA alpha 4 integrin chain. The molecule forms the heterodimer called VLA-4 with integrin beta 1 and another heterodimer with integrin beta 7 that binds fibronectin, VCAM-1, and MadCAM-1. CD49d is expressed on monocytes, lymphocytes, thymocytes, NK cells, B cells, and T cells. It is involved in hematopoietic stem cell differentiation, cell migration, and cell activation. Its absence on Foxp3+ cells make it a useful marker to isolate Treg cell populations.

Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: 9F10
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR