|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | IL7R |
Antigen Name | Interleukin 7 Receptor |
Reactivity | Mouse |
Conjugation | Biotin |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | A7R34 |
Host | Rat |
Isotype | IgG2a kappa |
Flow Cytometry | Yes |
Antigen Synonyms | CDW127, IL-7R-alpha, IL7RA, ILRA, CD127 |
Concentration | 0.5 mg/ml |
Cross Adsorption | No |
Purification | purified utilizing affinity chromatography and unreacted dye was removed from the product. |
Form | liquid |
Storage Buffer | Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2. |
Storage Temperature | The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze. |
Shipping Temperature | -20°C |