Anti-KIT Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: YB5.B8]

Supplier: ProSci
76-904
10765-004EA 601.93 USD
10765-004
Anti-KIT Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: YB5.B8]
Antibodies
The YB5.B8 monoclonal antibody specifically binds with human CD117 (c-kit), the 145 kDA receptor for stem cell factor (SCF) or steel factor. This tyrosine kinase protein is expressed mainly on hematopoietic progenitor, mast cells, acute myeloid leukemia cells (AML). CD117 activation promotes proliferation and cell differentiation on these cell subtypes. A lack or defect of SCF can lead to a decrease in number of hematopoietic progenitor cells and severe anemia.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: YB5.B8
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR