Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]

Supplier: Prosci
76-903
The ACK2 monoclonal antibody specifically reacts with mouse CD117 (c-Kit receptor), a 145 kDa transmembrane tyrosine-kinase receptor encoded by the Kit gene. The c-Kit receptor, also known as stem cell factor receptor, is expressed on hematopoietic progenitor cells in adult bone marrow, in progenitors of erythroid and myeloid lineages, and precursors of B and T cells. CD117 enhances the proliferation and the differentiation of the hematopoietic progenitor cells and seems to enhance the development of T cells, as the c-Kit receptor and its ligand are expressed by the thymus.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: ACK2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 µg 76-903 10765-002 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol KIT
Antigen Name Tyrosine-protein kinase Kit
Reactivity Mouse
Conjugation APC (Allophycocyanin)
Clonality Monoclonal
Antibody Type Primary
Clone ACK2
Host Rat
Isotype IgG2b kappa
Flow Cytometry Yes
Antigen Synonyms CD117, Ssm, SCO1, Fdc, Gsfsco5, SOW3, Bs, Tr-kit, SCO5, c-KIT, W, Gsfsco1, Gsfsow3
Concentration 0.2 mg/ml
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty