Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGA4 Mouse Monoclonal Antibody [clone: 9F10]

Supplier: Prosci
76-799
The 9F10 monoclonal antibody specifically reacts with human CD49d, the 150 kDA alpha 4 integrin chain. The molecule forms the heterodimer called VLA-4 with integrin beta 1 and another heterodimer with integrin beta 7 that binds fibronectin, VCAM-1, and MadCAM-1. CD49d is expressed on monocytes, lymphocytes, thymocytes, NK cells, B cells, and T cells. It is involved in hematopoietic stem cell differentiation, cell migration, and cell activation. Its absence on Foxp3+ cells make it a useful marker to isolate Treg cell populations.

Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: 9F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 76-799 10764-866 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ITGA4
Antigen Name Integrin Alpha 4
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 9F10
Host Mouse
Isotype IgG1 kappa
ImmunoChemistry Yes
Flow Cytometry Yes
Antigen Synonyms Integrin alpha 4, Integrin alpha-4, CD49D, IA4, CD49 antigen-like family member D
Concentration 0.5 mg/ml
Cross Adsorption No
Purification purified utilizing affinity chromatography.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty