Anti-ITGA2B Rat Monoclonal Antibody [clone: MWReg30]

Supplier: ProSci
76-170
10766-280EA 683.4 USD
10766-280
Anti-ITGA2B Rat Monoclonal Antibody [clone: MWReg30]
Antibodies
The MWReg30 monoclonal antibody specifically reacts with mouse CD41, a transmembrane glycoprotein also known as integrin alpha 2b GPIIb. It is expressed on hematopoietic progenitors, megakaryocytes, and platelets. It forms a receptor with CD61 (integrin beta 3) that binds adhesion molecules such as fibrinogen, fibronectin, von Willebrand factor, and thrombin. Defects or absence of CD41 has been found to lead to coagulation disorders. The expression profile of CD150+,CD48-, and CD41- can be used to identify hematopoietic stem cells.

Type: Primary
Antigen: Itga2b
Clonality: Monoclonal
Clone: MWReg30
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: Rat IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR