Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIP8]

Supplier: ProSci
76-169
10766-278EA 340.73 USD
10766-278
Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIP8]
Antibodies
The HIP8 monoclonal antibody specifically reacts with human CD41 (GPIIb), a subunit of the calcium-dependent complex with CD61 (IIIa). The complex is expressed on megakaryocytes and platelets. The HIP8 antibody blocks the aggregation of platelets and inhibits platelet activation.

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: HIP8
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR