Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]

Supplier: ProSci
76-463
10766-434EA 573.04 USD
10766-434
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI100
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR