Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AK-4]

Supplier: ProSci
76-198
10766-344EA 340.73 USD
10766-344
Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AK-4]
Antibodies
The AK-4 monoclonal antibody specifically reacts with human CD62P (P-Selectin), a 140 kDA type I transmembrane glycoprotein also known as platelet activation-dependent granule membrane protein (PADGEM). Upon activation it is quickly transported to the plasma membrane from the alpha-granules of platelets and WEibel-Palade bodies of endothelial cells. It is essential to the functions of cell adhesion during inflammatory reactions and in the interaction of platelets with monocytes and neutrophils.

Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: AK-4
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR