Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 30-F11]

Supplier: ProSci
76-531
10766-454EA 448.21 USD
10766-454
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 30-F11]
Antibodies
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PE-Cy7
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR