Anti-PECAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM59]

Supplier: ProSci
76-166
10766-272EA 413.33 USD
10766-272
Anti-PECAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM59]
Antibodies
The WM59 monoclonal antibody specifically reacts with human CD31, a 130-140 kDA type I transmembrane glycoprotein also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) or Endocam. CD31 is reported to bind to CD38 and is expressed on platelets, monocytes, granulocytes, and endothelial cells. It plays a role in angiogenesis, wound healing, cellular migration, and signal transduction.

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: WM59
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR