Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2]

Supplier: ProSci
76-430
10764-516EA 491.73 USD
10764-516
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2]
Antibodies
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 15.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR