Anti-IL2RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BC96]

Supplier: ProSci
76-505
10764-602EA 345.65 USD
10764-602
Anti-IL2RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BC96]
Antibodies
The BC96 monoclonal antibody specifically reacts with the 55 kDa type I transmembrane glycoprotein known as the interleukin-2 receptor alpha (IL-2R alpha, also known as CD25). CD25 is expressed by the early progenitors of T and B lymphocytes lineage, and by activated mature T and B lymphocytes. CD25 is a low affinity interleukin-2 receptor, but its association with the IL-2 receptor beta chain (CD122) and the common gamma chain (CD 132) results in a high affinity IL-2R complex. CD25 plays an important role in B and T cell proliferation, differentiation, and activation.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: BC96
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR