Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5E10]

Supplier: ProSci
76-068
10766-076EA 683.4 USD
10766-076
Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5E10]
Antibodies
The 5E10 monoclonal antibody specifically reacts with human CD90, also known as thymus cell antigen-1 (Thy-1). CD90 is a 25-37 kDA GPI-anchored protein is the smallest member of the Ig superfamily. CD90 is expressed on thymocytes, neurons, mesenchymal stem cells, hematopoietic stem cells, NK cells, and follicular dendritic cells and plays a role in inflammation, metastasis, apoptosis, and nerve regeneration. The 5E10 antibody can be used for the enriching high proliferative potential colony-forming cells (HIPP-CFC).

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: 5E10
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR