Anti-TNFSF4 Rat Monoclonal Antibody (Biotin) [clone: RM134L]

Supplier: ProSci
76-942
10765-072EA 297.25 USD
10765-072
Anti-TNFSF4 Rat Monoclonal Antibody (Biotin) [clone: RM134L]
Antibodies
The RM134L monoclonal antibody specifically reacts with mouse CD252, a TNF/NGF superfamily member present on activated B lymphocytes and antigen-presenting cells. On activated B cells it enhances immunoglobulin secretion and cell proliferation. CD252 is also known as the OX-40 ligand and interacts with the OX-40 antigen found on the surface of activated T cells. The RML134L is reported to block the costimulatory activity of OX-40L.

Type: Primary
Antigen: Tnfsf4
Clonality: Monoclonal
Clone: RM134L
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR