Anti-KDR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: Avas12a1]

Supplier: ProSci
76-992
10765-138EA 848.52 USD
10765-138
Anti-KDR Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: Avas12a1]
Antibodies
The Avas12a1 monoclonal antibody specifically reacts with mouse CD309, also known as fetal liver kinase-1 (Flk-1) or the vascular endothelial growth factor receptor 2 (VEGFR2). CD309 is a receptor for VEGF and VEGFC and is expressed in endothelial cells in embryonic and adult tissue and a requirement for the development of vascular endothelial and hematopoietic cells.

Type: Primary
Antigen: Kdr
Clonality: Monoclonal
Clone: Avas12a1
Conjugation: PE
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR