Anti-TNFRSF18 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DTA-1]

Supplier: ProSci
76-988
10765-130EA 345.65 USD
10765-130
Anti-TNFRSF18 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DTA-1]
Antibodies
Type: Primary
Antigen: Tnfrsf18
Clonality: Monoclonal
Clone: DTA-1
Conjugation: PE
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR