Anti-CD3E Mouse Monoclonal Antibody [clone: Hit3a]

Supplier: ProSci
76-213
10765-340EA 297.25 USD
10765-340
Anti-CD3E Mouse Monoclonal Antibody [clone: Hit3a]
Antibodies
The Hit3a monoclonal antibody specifically reacts with the Epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.The HIT3a antibody cross-links with the TCR complex, initiating the cellular activation and proliferation, but it cannot be used for intracellular CD3 staining, as it is able to stain only the surface CD3 complex.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: Hit3a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR