Anti-ENG Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MJ7/18]

Supplier: ProSci
76-828
10764-908EA 535.12 USD
10764-908
Anti-ENG Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MJ7/18]
Antibodies
The MJ7/18 monoclonal antibody specifically reacts with mouse CD105 (Endoglin), a 90kDA homodimeric glycoprotein expressed on vascular endothelial cells. CD105 is a marker for tumor angiogenesis research by identifying proliferating endothelium. It is also suggested to be involved in embryonic angiogenesis and cellular adhesion.

Type: Primary
Antigen: Eng
Clonality: Monoclonal
Clone: MJ7/18
Conjugation: PE
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR