Anti-Fcgr3 Rat Monoclonal Antibody [clone: 2.4G2]

Supplier: ProSci
76-573
10765-520EA 848.52 USD
10765-520
Anti-Fcgr3 Rat Monoclonal Antibody [clone: 2.4G2]
Antibodies
The 2.4G2 monoclonal antibody specifically reacts with an epitope on the extracellular domain of the mouse CD16 (Fc gamma III) and CD 32 (Fc gamma II). CD16 and CD32 are low affinity receptors for the IgG Fc domain and are expressed by B lymphocytes, NK cells, kupffer cells, mast cells, monocytes, macrophages, granulocytes, immature thymocytes, neutrophils, and some activated mature T cells.The 2.4G2 antibody blocks the binding of immunoglobulins to CD16 and CD32, and possibly to Fc gamma I receptor.

Type: Primary
Antigen: Fcgr3
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR