Anti-CD8B1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: H35-17.2]

Supplier: ProSci
76-721
10764-814EA 448.21 USD
10764-814
Anti-CD8B1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: H35-17.2]
Antibodies
The H35-17.2 monoclonal antibody specifically reacts the mouse cd8 beta, a 32 kDa chain also known as Ly-3 and Lyt-3. CD8 is a T cell surface antigen co-receptor that interacts with MHC class I molecules on antigen presenting cells. It is expressed as a heterodimer with the alpha cd8 chain. The H35-17.2 antibody blocks T cell-mediated cytolysis of allogeneic lymphoma cells.

Type: Primary
Antigen: Cd8b1
Clonality: Monoclonal
Clone: H35-17.2
Conjugation: APC
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR