Anti-CFTR Rabbit Polyclonal Antibody

Supplier: ProSci
7717_S
10752-174EA 187.23 USD
10752-174
Anti-CFTR Rabbit Polyclonal Antibody
Antibodies
The cystic fibrosis transmembrane conductance regulator (CFTR) protein is a member of the ATP-binding cassette (ABC) transporter superfamily, and a member of the MRP subfamily that is involved in multi-drug resistance. CFTR functions as a chloride channel and controls the regulation of other transport pathways. Mutations in this gene are associated with the autosomal recessive disorder cystic fibrosis, the most common, fatal, inherited disease of caucasian populations.

Type: Primary
Antigen: CFTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR