Anti-MKRN3 Rabbit Polyclonal Antibody

Supplier: ProSci
8145_S
10749-104EA 185.24 USD
10749-104
Anti-MKRN3 Rabbit Polyclonal Antibody
Antibodies
The Makorin ring finger 3 (MKRN3) protein contains a RING (C3HC4) zinc finger motif and several C3H zinc finger motifs. The MKRN3 gene is intronless and imprinted, with expression only from the paternal allele. Disruption of the imprinting at this locus may contribute to Prader-Willi syndrome , but a deletion of the gene does not. A deficiency of MKRN3 has been shown to cause central precocious puberty in humans.

Type: Primary
Antigen: MKRN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR