Anti-P2RX7 Rabbit Polyclonal Antibody

Supplier: ProSci
8015_S
10749-004EA 185.24 USD
10749-004
Anti-P2RX7 Rabbit Polyclonal Antibody
Antibodies
The purinergic receptor P2X ligand-gated ion channel 7 (P2RX7) belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Recent studies have suggested that P2RX7 may play a key role in immune-mediated diseases such as rheumatoid arthritis as well as neuropsychiatric disorders.

Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR