Anti-WDR61 Rabbit Polyclonal Antibody

Supplier: ProSci
7985_S
10748-988EA 187.23 USD
10748-988
Anti-WDR61 Rabbit Polyclonal Antibody
Antibodies
WDR61 (WD-repeat-containing protein 61), also known as SKI8 or REC14, is a subunit of the human PAF and SKI complexes, which function in transcriptional regulation and are involved in events downstream of RNA synthesis, such as RNA surveillance. PAF1C is implicated in regulation of development and maintenance of embryonic stem cell pluripotency. Component of the SKI complex which is thought to be involved in exosome-mediated RNA decay and associates with transcriptionally active genes in a manner dependent on PAF1C.

Type: Primary
Antigen: WDR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR