Anti-NDST2 Rabbit Polyclonal Antibody

Supplier: ProSci
7869_S
10748-962EA 185.24 USD
10748-962
Anti-NDST2 Rabbit Polyclonal Antibody
Antibodies
The N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2 (NDST2) protein, also known as N-heparan sulfate sulfotransferase 2, and the highly related protein NDST1 have key roles in the sulfation of heparan sulfate proteoglycans, molecules that are present of cell surfaces and the extracellular matrix. NDST1 and NDST2 are dispensable for mesodermal differentiation into osteoblasts but necessary for induction of adipocytes and neural cells.

Type: Primary
Antigen: NDST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR